ENA333557844 Enamine

3-[4-(propan-2-yl)phenyl]butanoic acid

  • MDL number MFCD01549654
  • Cas Number : 53086-50-3
  • Molecular Weight : 206.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81993
Availability details for
Quantity
  • Availability
  • Add to Cart