ENA333557850 Enamine

2-(2-fluorophenyl)-5-methyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07379312
  • Cas Number : 61152-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82004
Availability details for
Quantity
  • Availability
  • Add to Cart