ENA333557852 Enamine

1-(thiophen-2-yl)cyclohexane-1-carboxylic acid

  • MDL number MFCD03426862
  • Cas Number : 100058-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-147
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82008
Availability details for
Quantity
  • Availability
  • Add to Cart