ENA333557858 Enamine

1-methoxyhexan-2-one

  • MDL number MFCD16807848
  • Cas Number : 57134-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82014
Availability details for
Quantity
  • Availability
  • Add to Cart