ENA333557859 Enamine

1-(2-bromoethoxy)-3-methanesulfonylbenzene

  • MDL number MFCD12066170
  • Cas Number : 1157773-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82015
Availability details for
Quantity
  • Availability
  • Add to Cart