ENA333557865 Enamine

(2-chlorophenyl)(4-ethylphenyl)methanone

  • MDL number MFCD09801582
  • Cas Number : 82520-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82023

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82023
Availability details for
Quantity
  • Availability
  • Add to Cart