ENA333557867 Enamine

5-chloro-3-(3-phenoxyphenyl)-1,2,4-oxadiazole

  • MDL number MFCD12845386
  • Cas Number : 1183803-32-8
  • Molecular Weight : 272.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82025
Availability details for
Quantity
  • Availability
  • Add to Cart