ENA333557879 Enamine

2-cyclohexyl-1-(pyrimidin-2-yl)ethan-1-amine

  • MDL number MFCD16822528
  • Cas Number : 1250770-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77692
Availability details for
Quantity
  • Availability
  • Add to Cart