ENA333557886 Enamine

4-(hydroxymethyl)-1-methyl-5-(thiophen-2-yl)pyrrolidin-2-one

  • MDL number MFCD19686430
  • Cas Number : 1333524-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82048
Availability details for
Quantity
  • Availability
  • Add to Cart