ENA333557887 Enamine

1-[2-(pyridin-4-yl)ethyl]-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD03931550
  • Cas Number : 892149-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82049
Availability details for
Quantity
  • Availability
  • Add to Cart