ENA333557888 Enamine

8-fluoro-1-methyl-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-2-one

  • MDL number MFCD19686431
  • Cas Number : 1333665-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82050

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82050
Availability details for
Quantity
  • Availability
  • Add to Cart