ENA333557892 Enamine

2-hydroxy-2-[3-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD14601971
  • Cas Number : 3259-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82054

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82054
Availability details for
Quantity
  • Availability
  • Add to Cart