ENA333557898 Enamine

2-(thiophen-2-yl)propan-1-amine

  • MDL number MFCD01859703
  • Cas Number : 337471-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82062
Availability details for
Quantity
  • Availability
  • Add to Cart