ENA333557905 Enamine

1-(1-chloropropyl)-2,4-difluorobenzene

  • MDL number MFCD12788886
  • Cas Number : 1251280-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-82069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82069
Availability details for
Quantity
  • Availability
  • Add to Cart