ENA333557909 Enamine

5-chloro-2-[(2,2,2-trifluoroethyl)sulfanyl]aniline

  • MDL number MFCD11128871
  • Cas Number : 1019350-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82073
Availability details for
Quantity
  • Availability
  • Add to Cart