ENA333557910 Enamine

2-[(2,2,2-trifluoroethyl)sulfanyl]-5-(trifluoromethyl)aniline

  • MDL number MFCD11134427
  • Cas Number : 1094403-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82074
Availability details for
Quantity
  • Availability
  • Add to Cart