ENA333557911 Enamine

1-(4-amino-1H-pyrazol-1-yl)-3-[methoxy(methyl)amino]propan-2-ol

  • MDL number MFCD12095588
  • Cas Number : 1154213-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82075
Availability details for
Quantity
  • Availability
  • Add to Cart