ENA333557916 Enamine

3-(furan-2-yl)-1-(3-methylphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD08444932
  • Cas Number : 956515-98-3
  • Molecular Weight : 239.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82080
Availability details for
Quantity
  • Availability
  • Add to Cart