ENA333557918 Enamine

(3-chloro-6-hydrazinylpyridin-2-yl)methanol

  • MDL number MFCD17241249
  • Cas Number : 1334148-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82083
Availability details for
Quantity
  • Availability
  • Add to Cart