ENA333557920 Enamine

2-(5-fluoro-2-methylphenyl)-1H-1,3-benzodiazole

  • MDL number MFCD17619241
  • Cas Number : 1021397-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82085
Availability details for
Quantity
  • Availability
  • Add to Cart