ENA333557923 Enamine

1-(pyridin-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD01692454
  • Cas Number : 28465-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-76
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82089
Availability details for
Quantity
  • Availability
  • Add to Cart