ENA333557926 Enamine

4-{4-[(dimethylamino)methyl]piperidin-1-yl}aniline

  • MDL number MFCD16251975
  • Cas Number : 1224932-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82092
Availability details for
Quantity
  • Availability
  • Add to Cart