ENA333557931 Enamine

3-cyclopropylcyclohexan-1-one

  • MDL number MFCD19686436
  • Cas Number : 59939-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82106
Availability details for
Quantity
  • Availability
  • Add to Cart