ENA333557942 Enamine

2-(naphthalen-1-yl)-1H-1,3-benzodiazole

  • MDL number MFCD01564700
  • Cas Number : 2562-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82125
Availability details for
Quantity
  • Availability
  • Add to Cart