ENA333557944 Enamine

butyl(1-phenylethyl)amine hydrochloride

  • MDL number MFCD02671074
  • Cas Number : 71475-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82127
Availability details for
Quantity
  • Availability
  • Add to Cart