ENA333557951 Enamine

1-benzyl-N,2-dimethylpiperidin-4-amine

  • MDL number MFCD14692666
  • Cas Number : 1248088-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82135

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82135
Availability details for
Quantity
  • Availability
  • Add to Cart