ENA333557953 Enamine

5-methyl-4-phenyl-2-(pyrrolidin-2-yl)-1H-imidazole

  • MDL number MFCD19686440
  • Cas Number : 1153103-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82141
Availability details for
Quantity
  • Availability
  • Add to Cart