ENA333557957 Enamine

N,N-dimethyl-1,2,3,4-tetrahydroquinoline-7-sulfonamide

  • MDL number MFCD18914651
  • Cas Number : 1307240-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82148
Availability details for
Quantity
  • Availability
  • Add to Cart