ENA333557962 Enamine

3-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid

  • MDL number MFCD19686441
  • Cas Number : 64263-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82158

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82158
Availability details for
Quantity
  • Availability
  • Add to Cart