ENA333557965 Enamine

5-(ethoxycarbonyl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00722725
  • Cas Number : 74936-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82165
Availability details for
Quantity
  • Availability
  • Add to Cart