ENA333557968 Enamine

4-{[5-(trifluoromethyl)pyridin-2-yl]amino}butanoic acid

  • MDL number MFCD08445188
  • Cas Number : 923121-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82168
Availability details for
Quantity
  • Availability
  • Add to Cart