ENA333557971 Enamine

2-(1-chloroethyl)-1,4-dimethylbenzene

  • MDL number MFCD11186460
  • Cas Number : 57527-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82171
Availability details for
Quantity
  • Availability
  • Add to Cart