ENA333557973 Enamine

N,2-dihydroxy-3-methylbenzamide

  • MDL number MFCD00045720
  • Cas Number : 26071-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82173
Availability details for
Quantity
  • Availability
  • Add to Cart