ENA333557977 Enamine

2-carbamoylacetic acid

  • MDL number MFCD00154605
  • Cas Number : 2345-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-113
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82179
Availability details for
Quantity
  • Availability
  • Add to Cart