ENA333557980 Enamine

1-(oxan-4-yl)-1,4-diazepane

  • MDL number MFCD12825049
  • Cas Number : 1184240-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82183
Availability details for
Quantity
  • Availability
  • Add to Cart