ENA333557983 Enamine

2-methyl-3-[4-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD16077273
  • Cas Number : 1192772-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82186
Availability details for
Quantity
  • Availability
  • Add to Cart