ENA333557986 Enamine

ethyl 2-(cyclopropylamino)-1,3-thiazole-4-carboxylate

  • MDL number MFCD19686442
  • Cas Number : 126534-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82195
Availability details for
Quantity
  • Availability
  • Add to Cart