ENA333557996 Enamine

1-(3-cyanophenyl)-3-(prop-2-yn-1-yl)urea

  • MDL number MFCD14631636
  • Cas Number : 1249604-40-7
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-82210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82210
Availability details for
Quantity
  • Availability
  • Add to Cart