ENA333558008 Enamine

2-bromo-1-[3-(difluoromethoxy)phenyl]ethan-1-one

  • MDL number MFCD09259003
  • Cas Number : 443914-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82227

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82227
Availability details for
Quantity
  • Availability
  • Add to Cart