ENA333558009 Enamine

methyl[1-(pyridin-4-yl)propyl]amine

  • MDL number MFCD12154260
  • Cas Number : 1184131-82-5
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82229
Availability details for
Quantity
  • Availability
  • Add to Cart