ENA333558013 Enamine

3-(3-bromopropoxy)oxolane

  • MDL number MFCD18364738
  • Cas Number : 1334146-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82233
Availability details for
Quantity
  • Availability
  • Add to Cart