ENA333558210 Enamine

methyl[(2-phenyl-2H-1,2,3-triazol-4-yl)methyl]amine hydrochloride

  • MDL number MFCD19982394
  • Cas Number : 1334149-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82403
Availability details for
Quantity
  • Availability
  • Add to Cart