ENA333558235 Enamine

2-amino-5-chloro-3-(2-ethoxy-5-fluorophenyl)-3H-indol-3-ol

  • MDL number MFCD19982402
  • Cas Number : 1299311-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82511
Availability details for
Quantity
  • Availability
  • Add to Cart