ENA333558253 Enamine

2-(benzylamino)-3,3,3-trifluoropropan-1-ol

  • MDL number MFCD19686511
  • Cas Number : 126536-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82585

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-82585
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart