ENA333558258 Enamine

4-[4-(difluoromethoxy)phenyl]oxan-4-amine hydrochloride

  • MDL number MFCD19982411
  • Cas Number : 1334147-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82609
Availability details for
Quantity
  • Availability
  • Add to Cart