ENA333558263 Enamine

2-(1-aminocyclohexyl)-6-cyclopropyl-3,4-dihydropyrimidin-4-one hydrochloride

  • MDL number MFCD19982426
  • Cas Number : 1334147-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82647
Availability details for
Quantity
  • Availability
  • Add to Cart