ENA333558266 Enamine

9-{[(tert-butoxy)carbonyl]amino}bicyclo[3.3.1]nonane-3-carboxylic acid

  • MDL number MFCD19686518
  • Cas Number : 1334148-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82673
Availability details for
Quantity
  • Availability
  • Add to Cart