ENA333558296 Enamine

1-cyclopropyl-5-(propan-2-yl)-1H-imidazole-2-thiol

  • MDL number MFCD19982434
  • Cas Number : 1334149-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82735

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82735
Availability details for
Quantity
  • Availability
  • Add to Cart