ENA333558297 Enamine

1,5-bis(propan-2-yl)-1H-imidazole-2-thiol

  • MDL number MFCD19982435
  • Cas Number : 1334146-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82736
Availability details for
Quantity
  • Availability
  • Add to Cart