ENA333558306 Enamine

5-fluoropyridine-2-sulfonyl fluoride

  • MDL number MFCD19686554
  • Cas Number : 1210867-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82821
Availability details for
Quantity
  • Availability
  • Add to Cart